missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N’,N’-Tetramethylethane-1,2-diamine Dihydrochloride (1,2-Bis(dimethylamino)ethane Dihydrochloride, N,N,N’,N’-Tetramethylethylenediamine Dihydrochloride), Mikromol
Description
N,N,N’,N’-Tetramethylethane-1,2-diamine Dihydrochloride (1,2-Bis(dimethylamino)ethane Dihydrochloride, N,N,N’,N’-Tetramethylethylenediamine Dihydrochloride), Mikromol
Specifications
Specifications
| Chemical Name or Material | N,N,N',N'-Tetramethylethane-1,2-diamine Dihydrochloride |
| CAS | 7677-21-6 |
| Analyte or Component Names | N,N,N',N'-Tetramethylethane-1,2-diamine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H16 N2 . 2 Cl H |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 1,2-Ethanediamine, N1,N1,N2,N2-tetramethyl-, hydrochloride (1:2), 1,2-Ethanediamine, N,N,N',N'-tetramethyl-, dihydrochloride (9CI), Ethylenediamine, N,N,N',N'-tetramethyl-, dihydrochloride (8CI), N,N,N',N'-Tetramethylethylenediamine dihydrochloride, Potassium Clavulanate Impurity J |
| InChI Formula | InChI=1S/C6H16N2.2ClH/c1-7(2)5-6-8(3)4;;/h5-6H2,1-4H3;2*1H |
| Solution Type | Neat |
| SMILES | Cl.Cl.CN(C)CCN(C)C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?