missing translation for 'onlineSavingsMsg'
Learn More

N,N,N-Trimethyl-1-phenylmethanaminium bromide, 97%, BLD Pharm™

Product Code. 31005770
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31005770 25 g 25g
30997182 1 kg 1kg
30998117 500 g 500g
30998596 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31005770 Supplier BLD Pharm Supplier No. BD11635325g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5350-41-4
Molecular Formula C10H16BrN
MDL Number MFCD00011780
Quantity 25 g
SMILES C[N+](C)(C)CC1=CC=CC=C1.[Br-]
Molecular Weight (g/mol) 230.15
Percent Purity 97%
Chemical Name or Material N,N,N-Trimethyl-1-phenylmethanaminium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.