missing translation for 'onlineSavingsMsg'
Learn More

N,N'-(Octane-1,8-diyl)bis(2,2-dichloroacetamide), 95%, BLD Pharm™

Product Code. 30783514
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
Unit Size:
100mg
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30783514 100 mg 100mg
30778124 250 mg 250mg
30768618 5 g 5g
30782820 1 g 1g
30774968 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30783514 Supplier BLD Pharm Supplier No. BD142371100mg
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1477-57-2
Molecular Formula C12H20Cl4N2O2
MDL Number MFCD00000841
Quantity 100 mg
SMILES O=C(NCCCCCCCCNC(C(Cl)Cl)=O)C(Cl)Cl
Molecular Weight (g/mol) 366.11
Percent Purity 95%
Chemical Name or Material N,N'-(Octane-1,8-diyl)bis(2,2-dichloroacetamide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.