missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso Fluoxetine, TRC

Product Code. 17992755
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992755 250 mg 250mg
17921445 500 mg 500mg
17942205 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992755 Supplier Toronto Research Chemicals Supplier No. TRCN171730250MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150494-06-7
Molecular Formula C17 H17 F3 N2 O2
Molecular Weight (g/mol) 338.3243
Synonym Benzenepropanamine, N-methyl-N-nitroso-γ-[4-(trifluoromethyl)phenoxy]-, N-Methyl-N-nitroso-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine, N-Nitroso Fluoxetine, N-Nitrosofluoxetine
IUPAC Name N-methyl-N-[3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]nitrous amide
SMILES CN(CCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2)N=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H17F3N2O2/c1-22(21-23)12-11-16(13-5-3-2-4-6-13)24-15-9-7-14(8-10-15)17(18,19)20/h2-10,16H,11-12H2,1H3
Formula Weight 338.1242
Percent Purity >95%
Chemical Name or Material N-Nitrosofluoxetine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.