missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N-ethylaniline, TRC

Product Code. 30041194
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041194 100 mg 100mg
30010634 50 mg 50mg
18072050 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041194 Supplier Toronto Research Chemicals Supplier No. TRCN525875100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-64-6
Molecular Formula C8 H10 N2 O
Molecular Weight (g/mol) 150.18
Synonym Aniline, N-ethyl-N-nitroso- (6CI,7CI,8CI), N-Ethyl-N-nitrosobenzenamine, N-Ethyl-N-nitrosoaniline, N-Ethyl-N-phenylnitrosamine, N-Nitroso-N-ethylaniline, N-Nitrosoethylphenylamine, NSC 405070, N-Nitroso-N-ethyl-N-phenylamine, NEPhA
IUPAC Name N-ethyl-N-phenylnitrous amide
SMILES CCN(N=O)c1ccccc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H10N2O/c1-2-10(9-11)8-6-4-3-5-7-8/h3-7H,2H2,1H3
Formula Weight 150.0793
Percent Purity >95%
Chemical Name or Material N-Nitroso-N-ethylaniline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.