Learn More
N-Nitroso-N-methyl-N-tetradecylamine, TRC
Product Code. 30138061
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138061 50 mg 50mg
30010682 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138061 Supplier Toronto Research Chemicals Supplier No. TRCN52996050MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75881-20-8
Molecular Formula C15H32N2O
Molecular Weight (g/mol) 256.43
Synonym N-Methyl-N-nitroso-1-tetradecanamine (ACI), N-Nitroso-N-methyltetradecylamine
SMILES CCCCCCCCCCCCCCN(C)N=O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C15H32N2O/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-17(2)16-18/h3-15H2,1-2H3
Formula Weight 256.2515
Percent Purity >95%
Chemical Name or Material N-Nitroso-N-methyl-N-tetradecylamine
Grade HPLC