missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N-methylaniline (100 ug/mL in methanol), TRC

Product Code. 30007844
Change view
Click to view available options
Quantity:
1 x 1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007844 1 x 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30007844 Supplier Toronto Research Chemicals Supplier No. TRCKIT29921X1ML
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 614-00-6
Molecular Formula C7 H8 N2 O
Quantity 1 x 1 mL
Synonym Aniline, N-methyl-N-nitroso- (6CI,8CI), N-Methyl-N-nitrosobenzenamine, Methylphenylnitrosamine, Methylphenylnitrosoamine, N-Methyl-N-nitrosoaniline, N-Methyl-N-phenylnitrosamine, N-Methyl-N-phenylnitrosoamine, N-Nitroso-N-methylaniline, N-Nitroso-N-methylphenylamine, NSC 137, Nitrosomethylphenylamine, Phenylmethylnitrosamine
InChI Formula InChI=1S/C7H8N2O/c1-9(8-10)7-5-3-2-4-6-7/h2-6H,1H3
SMILES CN(N=O)c1ccccc1
IUPAC Name N-methyl-N-phenylnitrous amide
Molecular Weight (g/mol) 136.15
Formula Weight 136.0637
Concentration or Composition (by Analyte or Components) N-Nitroso-N-methylaniline 100 μg/mL
Chemical Name or Material N-Nitroso-N-methylaniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.