missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N,N-di(3,5,5-trimethylhexyl)amine (>85%), TRC

Product Code. 17938014
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17938014 100 mg 100mg
17999644 25 mg 25mg
17938414 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17938014 Supplier Toronto Research Chemicals Supplier No. TRCN525800100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1207995-62-7
Molecular Formula C18 H38 N2 O
Molecular Weight (g/mol) 298.5071
Synonym 3,5,5-Trimethyl-N-nitroso-N-(3,5,5-trimethylhexyl)-1-hexanamine, 1-Hexanamine, 3,5,5-trimethyl-N-nitroso-N-(3,5,5-trimethylhexyl)-, N-Nitrosodiisononylamine
IUPAC Name N,N-bis(3,5,5-trimethylhexyl)nitrous amide
SMILES CC(CCN(CCC(C)CC(C)(C)C)N=O)CC(C)(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H38N2O/c1-15(13-17(3,4)5)9-11-20(19-21)12-10-16(2)14-18(6,7)8/h15-16H,9-14H2,1-8H3
Formula Weight 298.2984
Chemical Name or Material N-Nitrosodiisononylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.