missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosoatrazine, TRC

Product Code. 30135866
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135866 1 g 1g
30006091 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135866 Supplier Toronto Research Chemicals Supplier No. TRCN5248001G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56525-09-8
Molecular Formula C8 H13 Cl N6 O
Molecular Weight (g/mol) 244.68
Synonym 6-Chloro-N2-ethyl-N4-(1-methylethyl)-N2-nitroso-1,3,5-triazine-2,4-diamine, 6-Chloro-N-ethyl-N'-(1-methylethyl)-N-nitroso-1,3,5-triazine-2,4-diamine;
IUPAC Name N-[4-chloro-6-(propan-2-ylamino)-1,3,5-triazin-2-yl]-N-ethylnitrous amide
SMILES CCN(N=O)c1nc(Cl)nc(NC(C)C)n1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H13ClN6O/c1-4-15(14-16)8-12-6(9)11-7(13-8)10-5(2)3/h5H,4H2,1-3H3,(H,10,11,12,13)
Formula Weight 244.0839
Percent Purity >95%
Chemical Name or Material N-Nitrosoatrazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.