missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiclofenac, TRC

Product Code. 30041242
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041242 10 mg 10mg
30041375 50 mg 50mg
30010886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041242 Supplier Toronto Research Chemicals Supplier No. TRCN13743010MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66505-80-4
Molecular Formula C14H10Cl2N2O3
Molecular Weight (g/mol) 325.15
Synonym Benzeneacetic acid, 2-[(2,6-dichlorophenyl)nitrosoamino]-, 2-[(2,6-Dichlorophenyl)nitrosoamino]benzeneacetic acid, N-Nitrosodiclofenac, NCX 284, 2-[(2,6-Dichlorophenyl)nitrosoamino]benzeneacetic Acid, NCX 284
IUPAC Name 2-[2-(2,6-dichloro-N-nitrosoanilino)phenyl]acetic acid
SMILES OC(=O)Cc1ccccc1N(N=O)c2c(Cl)cccc2Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H10Cl2N2O3/c15-10-5-3-6-11(16)14(10)18(17-21)12-7-2-1-4-9(12)8-13(19)20/h1-7H,8H2,(H,19,20)
Formula Weight 324.0068
Percent Purity >95%
Chemical Name or Material N-Nitrosodiclofenac
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.