missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodimethyl-1,1,1-d3-amine, CDN

Product Code. 30325371
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Product Code. Quantity unitSize
30325371 0.05 g 0.05g
30320182 0.1 g 0.1g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30325371

Brand: CDN CDND41080.05G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Nitroso-dimethylamine methyl-d3
CAS 32745-07-6
Molecular Formula C2 2H3 H3 N2 O
Quantity 0.05 g
Synonym Methan-d3-amine, N-methyl-N-nitroso- (9 CI), Dimethylamine-1,1,1-d3, N-nitroso- (8 CI), N-Nitroso-dimethylamine (methyl-D3), N-Nitroso- N-methyl-d3-methylamine
InChI Formula InChI=1 S/C2H6N2O/c1-4(2)3-5/h1-2H3/i1D3
SMILES [2 H]C([2 H])([2 H])N(C)N=O
IUPAC Name N-methyl-N-(trideuteriomethyl)nitrous amide
Molecular Weight (g/mol) 77.1
Formula Weight 77.0668 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.