missing translation for 'onlineSavingsMsg'
Learn More

N-Nitrosodiphenylamine, TRC

Product Code. 30005533
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005533 5 g 5g
17934535 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005533 Supplier Toronto Research Chemicals Supplier No. TRCN5257755G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-30-6
Molecular Formula C12 H10 N2 O
Molecular Weight (g/mol) 198.22
Synonym Diphenylamine, N-nitroso- (7CI,8CI), Diphenam N, Diphenylnitrosamine, N,N-Diphenyl-N-nitrosoamine, N,N-Diphenylnitrosamine, N-NITROSODIPHENYLAMINE, N-Nitroso-N-diphenylamine, N-Nitroso-N-phenylaniline, N-Nitroso-N-phenylbenzenamine, N-Nitrosodiphenylamine, N-Phenyl-N-nitrosoaniline, NSC 585, Nitrosodiphenylamine, Ortard, Redax, Retarder J, Sconoc, Vulcatard A, Vulkalent A, Vultrol, Benzenamine, N-nitroso-N-phenyl-
IUPAC Name N,N-diphenylnitrous amide
SMILES O=NN(c1ccccc1)c2ccccc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H10N2O/c15-13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H
Formula Weight 198.0793
Percent Purity >95%
Chemical Name or Material N-Nitroso-diphenylamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.