missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosoethylmethyl-d3-amine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-Nitrosoethylmethylamine-d3 (Major) |
| CAS | 69278-54-2 |
| Molecular Formula | C3D3H5N2O |
| Quantity | 0.01 g |
| Synonym | N-Methyl-N-nitrosoethanamine-d3, N-Nethyl-N-nitrosoethylamine-d3, Ethylmethylnitrosamine-d3, Methylethylnitrosamine-d3, Methylethylnitrosoamine-d3, N-Methyl-N-nitrosoethylamine-d3, N-Nitroso-N-methylethylamine-d3, N-Nitrosomethylethylamine-d3, NEMA-d3, Nitrosomethylethylamine-d3; |
| InChI Formula | InChI=1 S/C3H8N2O/c1-3-5(2)4-6/h3H2,1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])N(CC)N=O |
| IUPAC Name | N-ethyl-N-(trideuteriomethyl)nitrous amide |
| Molecular Weight (g/mol) | 91.13 |
| Formula Weight | 91.082 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction