missing translation for 'onlineSavingsMsg'
Learn More

N,O-Diacetate Isoacyclovir, TRC

Product Code. 30135680
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135680 10 mg 10mg
30136864 100 mg 100mg
30008653 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135680 Supplier Toronto Research Chemicals Supplier No. TRCD10310010MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91702-60-2
Molecular Formula C12 H15 N5 O5
Molecular Weight (g/mol) 309.278
IUPAC Name 2-[(2-acetamido-6-oxo-1H-purin-7-yl)methoxy]ethyl acetate
SMILES CC(=O)NC1=Nc2ncn(COCCOC(=O)C)c2C(=O)N1

Specifications

Quantity 10 mg
Synonym Acetamide, N-[7-[[2-(acetyloxy)ethoxy]methyl]-6,7-dihydro-6-oxo-1H-purin-2-yl]- (9CI, ACI), N-[7-[[2-(Acetyloxy)ethoxy]methyl]-6,7-dihydro-6-oxo-1H-purin-2-yl]acetamide
InChI Formula InChI=1S/C12H15N5O5/c1-7(18)14-12-15-10-9(11(20)16-12)17(5-13-10)6-21-3-4-22-8(2)19/h5H,3-4,6H2,1-2H3,(H2,14,15,16,18,20)
Formula Weight 309.107
Chemical Name or Material 2-[(2-acetamido-6-oxo-1H-purin-7-yl)methoxy]ethyl acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.