missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Octane-d18 (Reagent grade), CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | n-Octane (D18) |
| CAS | 17252-77-6 |
| Molecular Formula | C8 D18 |
| Quantity | 5 g |
| InChI Formula | InChI=1 S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-octadecadeuteriooctane |
| Molecular Weight (g/mol) | 132.34 |
| Formula Weight | 132.2538 g/mol |
| Percent Purity | 99 atom % D, min 99% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction