missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl gallate, 98+%

Product Code. 11302366
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302366 100 g 100g
11392356 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302366 Supplier Thermo Scientific Alfa Aesar Supplier No. L06081.22
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1034-01-1 | C15H22O5 | 282.336 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1034-01-1
Molecular Formula C15H22O5
Molecular Weight (g/mol) 282.336
MDL Number MFCD00002197
InChI Key NRPKURNSADTHLJ-UHFFFAOYSA-N
Synonym octyl gallate, n-octyl gallate, progallin o, stabilizer ga 8, n-octylgallate, gallic acid, octyl ester, gallic acid n-octyl ester, gallic acid octyl ester, benzoic acid, 3,4,5-trihydroxy-, octyl ester, oktylester kyseliny gallove
PubChem CID 61253
ChEBI CHEBI:83631
IUPAC Name octyl 3,4,5-trihydroxybenzoate
SMILES CCCCCCCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 96°C to 102°C
Quantity 100 g
Beilstein 2132305
Formula Weight 282.34
Percent Purity ≥98%
Chemical Name or Material n-Octyl gallate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.