missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Pentyl-2,2,3,3,4,4,5,5,5-d9 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Pentanol-2,2,3,3,4,4,5,5,5-D9 |
| CAS | 148587-12-6 |
| Molecular Formula | C5 D9 H3 O |
| Quantity | 0.25 g |
| Synonym | 1-Pentan-2,2,3,3,4,4,5,5,5-d9-ol (9 CI), 1-Pentanol-2,2,3,3,4,4,5,5,5-D9, 1-Pentanol-d9, n-Pentyl-2,2,3,3,4,4,5,5,5-d9 alcohol |
| InChI Formula | InChI=1 S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3/i1D3,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CO |
| IUPAC Name | 2,2,3,3,4,4,5,5,5-nonadeuteriopentan-1-ol |
| Molecular Weight (g/mol) | 97.204 |
| Formula Weight | 97.145 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction