missing translation for 'onlineSavingsMsg'
Learn More

n-Pentyl acetate, 99%

Product Code. 11333036
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333036 500 g 500g
11323036 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11333036 Supplier Thermo Scientific Alfa Aesar Supplier No. L10852.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 628-63-7 | C7H14O2 | 130.187 g/mol

n-Pentyl acetate is used as in studies of olfactory function and in studies of the psychosocial effects of odor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Pentyl acetate is used as in studies of olfactory function and in studies of the psychosocial effects of odor.

Solubility
Slightly soluble in water (2g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-63-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009500
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
Synonym amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
PubChem CID 12348
ChEBI CHEBI:87362
IUPAC Name pentyl acetate
SMILES CCCCCOC(=O)C

Specifications

Melting Point -100°C
Density 0.876
Boiling Point 142°C
Flash Point 24°C (75°F)
Odor Fruit-like
Refractive Index 1.403
Quantity 500 g
UN Number UN1104
Beilstein 1744753
Solubility Information Slightly soluble in water (2g/L).
Formula Weight 130.19
Percent Purity 99%
Chemical Name or Material n-Pentyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.