Learn More
n-Pentylcyclohexane, 98%
Product Code. 11409247
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409247 2 g 2g
11419247 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11409247 Supplier Thermo Scientific Alfa Aesar Supplier No. B21326.04
 Restricted Item
In Stock
Add to basket
CAS: 4292-92-6 | C11H22 | 154.297 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4292-92-6
Molecular Formula C11H22
Molecular Weight (g/mol) 154.297
MDL Number MFCD00019402
InChI Key HLTMUYBTNSVOFY-UHFFFAOYSA-N
Synonym n-amylcyclohexane, cyclohexane, pentyl, 1-cyclohexylpentane, n-pentylcyclohexane, amylcyclohexane, cyclohexane, n-pentyl, pentyl-cyclohexane, n-amyl cyclohexane, acmc-209jru, pentane, 1-cyclohexyl
PubChem CID 20284
IUPAC Name pentylcyclohexane
SMILES CCCCCC1CCCCC1
Specifications
Melting Point -58°C
Density 0.8
Boiling Point 202°C
Flash Point 63°C (145°F)
Odor Mild
Refractive Index 1.4445
Quantity 2 g
UN Number UN3082
Formula Weight 154.3
Percent Purity 98%
Chemical Name or Material n-Pentylcyclohexane