missing translation for 'onlineSavingsMsg'
Learn More

n-Pentylcyclohexane, 98%

Product Code. 11419247
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419247 10 g 10g
11409247 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419247 Supplier Thermo Scientific Alfa Aesar Supplier No. B21326.09
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 4292-92-6 | C11H22 | 154.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4292-92-6
Molecular Formula C11H22
Molecular Weight (g/mol) 154.297
MDL Number MFCD00019402
InChI Key HLTMUYBTNSVOFY-UHFFFAOYSA-N
Synonym n-amylcyclohexane, cyclohexane, pentyl, 1-cyclohexylpentane, n-pentylcyclohexane, amylcyclohexane, cyclohexane, n-pentyl, pentyl-cyclohexane, n-amyl cyclohexane, acmc-209jru, pentane, 1-cyclohexyl
PubChem CID 20284
IUPAC Name pentylcyclohexane
SMILES CCCCCC1CCCCC1

Specifications

Melting Point -58°C
Density 0.8
Boiling Point 202°C
Flash Point 63°C (145°F)
Odor Mild
Refractive Index 1.4445
Quantity 10 g
UN Number UN3082
Formula Weight 154.3
Percent Purity 98%
Chemical Name or Material n-Pentylcyclohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.