missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-p-phenylenediamine(p-Anilinoaniline), TRC

Product Code. 30339540
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339540 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339540 Supplier Toronto Research Chemicals Supplier No. TRCA663390100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Phenyl-1,4-phenylenediamine
CAS 101-54-2
Molecular Formula C12 H12 N2
Quantity 100 g
Synonym 1,4-Benzenediamine, N-phenyl- (9CI), Variamine Blue RT (6CI), p-Phenylenediamine, N-phenyl- (8CI), N1-Phenyl-1,4-benzenediamine, (4-(Phenylamino)phenyl)amine, 4-(N-Phenylamino)aniline, 4-(Phenylamino)aniline, 4-Amino-N-phenylaniline, 4-Aminodiphenylamine, 4-Anilinoaniline, 4-Anilinophenylamine, 4ADPA, Azoic Diazo Component 22, C.I. 76085, C.I. Developer 15, C.I. Oxidation Base 2, Diphenyl Black, Luxan Black R, N,4'-Bianiline, N-(4-Aminophenyl)aniline, N-Phenyl-1,4-benzenediamine, N-Phenyl-1,4-phenylenediamine, N-Phenyl-p-aminoaniline, N-Phenyl-p-phenylenediamine, NSC 3401, NSC 37074, Peltol BR, Peltol BR II, S 789, Semidin, Semidine, p-(Phenylamino)aniline, p-Aminodiphenylamine, p-Anilinoaniline, p-Semidine
InChI Formula InChI=1S/C12H12N2/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9,14H,13H2
SMILES Nc1ccc(Nc2ccccc2)cc1
IUPAC Name 4-N-phenylbenzene-1,4-diamine
Molecular Weight (g/mol) 184.24
Formula Weight 184.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.