missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylglycine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | N-Phenylglycine |
| CAS | 103-01-5 |
| Molecular Formula | C8H9NO2 |
| Quantity | 1 g |
| Synonym | (Phenylamino)acetic Acid, 2-(Phenylamino)acetic Acid, Anilinoacetic Acid, N-(Phenylamino)acetic Acid, NPG, NSC 83567 |
| InChI Formula | InChI=1S/C8H9NO2/c10-8(11)6-9-7-4-2-1-3-5-7/h1-5,9H,6H2,(H,10,11) |
| SMILES | OC(=O)CNc1ccccc1 |
| IUPAC Name | 2-anilinoacetic acid |
| Molecular Weight (g/mol) | 151.16 |
| Formula Weight | 151.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction