missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylthiourea, 97%

Product Code. 11406740
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406740 250 g 250g
11486730 10 g 10g
11496730 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406740 Supplier Thermo Scientific Alfa Aesar Supplier No. L06690.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 103-85-5 | C7H8N2S | 152.22 g/mol

N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

Solubility
Soluble in hot water and alcohol.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-85-5
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.22
MDL Number MFCD00004933
InChI Key FULZLIGZKMKICU-UHFFFAOYSA-N
Synonym n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
PubChem CID 676454
ChEBI CHEBI:46261
IUPAC Name phenylthiourea
SMILES NC(=S)NC1=CC=CC=C1

Specifications

Melting Point 149°C to 153°C
Odor Odorless
Quantity 250 g
UN Number UN2811
Beilstein 907309
Merck Index 14,7315
Solubility Information Soluble in hot water and alcohol.
Formula Weight 152.21
Percent Purity 97%
Chemical Name or Material N-Phenylthiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.