missing translation for 'onlineSavingsMsg'
Learn More

N-Propargyloxyphthalimide, 97%, Thermo Scientific™

Product Code. 15438899
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438899 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15438899 Supplier Thermo Scientific Alfa Aesar Supplier No. L09233.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4616-63-1
Melting Point 150°C to 152°C
Molecular Formula C11H7NO3
MDL Number MFCD00005890
Quantity 5 g
Beilstein 1530730
Synonym n-propargyloxy phthalimide, n-prop-2-ynyloxy phthalimide, 2-prop-2-yn-1-yloxy isoindoline-1,3-dione, n-propargyloxyphthalimide, 2-prop-2-yn-1-yloxy isoindole-1,3-dione, 2-prop-2-ynyloxybenzo c azoline-1,3-dione, acmc-20aka8, n-2-propynoxyphthalimide, n-2-propynyloxy phthalimide, phthalimide, n-2-propynyloxy
InChI Key HBGZBVPXPDNXOV-UHFFFAOYSA-N
SMILES C#CCON1C(=O)C2=CC=CC=C2C1=O
IUPAC Name 2-prop-2-ynoxyisoindole-1,3-dione
Molecular Weight (g/mol) 201.181
PubChem CID 78357
Formula Weight 201.18
Percent Purity 97%
Chemical Name or Material N-Propargyloxyphthalimide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.