Learn More
n-Propyl 4-aminobenzoate, 98%
Product Code. 11382856
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382856 50 g 50g
11372856 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11382856 Supplier Thermo Scientific Alfa Aesar Supplier No. L09066.18
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
CAS: 94-12-2 | C10H13NO2 | 179.22 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 94-12-2
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00017111
InChI Key NBFQYHKHPBMJJV-UHFFFAOYSA-N
Synonym risocaine, propaesin, propazyl, propesine, propylcain, raythesin, keloform p, propyl p-aminobenzoate, propesin, 4-propoxycarbonyl aniline
PubChem CID 7174
IUPAC Name propyl 4-aminobenzoate
SMILES CCCOC(=O)C1=CC=C(N)C=C1
Specifications
Melting Point 73°C to 75°C
Quantity 50 g
Beilstein 1211203
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material n-Propyl 4-aminobenzoate