missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl decanoate, 98%

Product Code. 11436247
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436247 50 g 50g
11446247 250 g 250g
11426247 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436247 Supplier Thermo Scientific Alfa Aesar Supplier No. B25159.18
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 30673-60-0 | C13H26O2 | 214.349 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30673-60-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
MDL Number MFCD00056329
InChI Key OVFMRFMJVFDSAA-UHFFFAOYSA-N
Synonym n-propyl decanoate, decanoic acid, propyl ester, propyl caprate, capric acid propyl ester, unii-s0he9grs1w, decanoic acid, propylester, s0he9grs1w, propyl caprate natural, propyl caprate decanoate, n-propyldecanoate
PubChem CID 121739
ChEBI CHEBI:86973
IUPAC Name propyl decanoate
SMILES CCCCCCCCCC(=O)OCCC

Specifications

Density 0.86
Boiling Point 254°C
Flash Point 115°C (239°F)
Quantity 50 g
Formula Weight 214.35
Percent Purity 98%
Chemical Name or Material n-Propyl decanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.