missing translation for 'onlineSavingsMsg'
Learn More

N-Propylsulfamide Potassium Salt, TRC

Product Code. 30426213
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426213 1 g 1g
30425445 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426213 Supplier Toronto Research Chemicals Supplier No. TRCP8384751G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1393813-41-6
Molecular Formula C3H10KN2O2S
Molecular Weight (g/mol) 177.29
Synonym Propylsulfamide potassium
IUPAC Name potassium propyl(sulfamoyl)amide
SMILES NS([N-]CCC)(=O)=O.[K+]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H9N2O2S.K/c1-2-3-5-8(4,6)7;/h2-3H2,1H3,(H2,4,6,7);/q-1;+1
Formula Weight 176
Chemical Name or Material N-Propylsulfamide Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.