Learn More
N-Succinimidyl Myristate, TRC
Product Code. 30424813
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424813 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30424813 Supplier Toronto Research Chemicals Supplier No. TRCS6908001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Succinimidyl Myristate
CAS 69888-86-4
Molecular Formula C18 H31 N O4
Quantity 1 g
InChI Formula InChI=1S/C18H31NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-18(22)23-19-16(20)14-15-17(19)21/h2-15H2,1H3
SMILES CCCCCCCCCCCCCC(=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) tetradecanoate
Molecular Weight (g/mol) 325.44
Formula Weight 325.2253