missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Tert-Butoxycarbonyl-3-Bromopropylamine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | N-Tert-Butoxycarbonyl-3-Bromopropylamine |
| CAS | 83948-53-2 |
| Molecular Formula | C8 H16 Br N O2 |
| Quantity | 1 g |
| Synonym | N-(3-Bromopropyl)Carbamic Acid 1,1-Dimethylethyl Ester, (3-Bromopropyl)Carbamic Acid 1,1-Dimethylethyl Ester, 1,1-Dimethylethyl (3-Bromopropyl)Carbamate, 1-[(Tert-Butoxycarbonyl)Amino]-3-Bromopropane, 3-(Boc-Amino)Propyl Bromide, 3-Bromo-N-(Tert-Butoxycarbonyl)Propylamine, 3-Bromopropylcarbamic Acid Tert-Butyl Ester, 3-[(Tert-Butoxycarbonyl)Amino]Propyl Bromide, N-(3-Bromopropyl)-N-[(1,1-Dimethylethoxy)Carbonyl]Amine, N-(3-Bromopropyl)Carbamic Acid Tert-Butyl Ester, N-Boc-3-Bromopropanamine, N-Boc-3-Bromopropylamine, Tert-Butyl N-(3-Bromopropyl)Carbamate |
| InChI Formula | InChI=1S/C8H16BrNO2/c1-8(2,3)12-7(11)10-6-4-5-9/h4-6H2,1-3H3,(H,10,11) |
| SMILES | Cc(C)(C)Oc(=O)Ncccbr |
| IUPAC Name | tert-butyl N-(3-bromopropyl)carbamate |
| Molecular Weight (g/mol) | 238.12 |
| Formula Weight | 237.0364 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction