missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-tert-Butoxycarbonyl 8-Methoxy Amoxapine-d8, TRC
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | N-tert-Butoxycarbonyl 8-Methoxy Amoxapine-d8 |
| Molecular Formula | C23 D8 H18 Cl N3 O4 |
| Quantity | 2.5 mg |
| Synonym | 4-(2-Chloro-8-Methoxydibenz[b,f][1,4]oxazepin-11-yl)-1-piperazinecarboxylic-d8 Acid 1,1-Dimethylethyl Ester |
| InChI Formula | InChI=1S/C23H26ClN3O4/c1-23(2,3)31-22(28)27-11-9-26(10-12-27)21-17-13-15(24)5-7-19(17)30-20-8-6-16(29-4)14-18(20)25-21/h5-8,13-14H,9-12H2,1-4H3/i9D2,10D2,11D2,12D2 |
| SMILES | [2H]C1([2H])N(C(=O)OC(C)(C)C)C([2H])([2H])C([2H])([2H])N(C2=Nc3cc(OC)ccc3Oc4ccc(Cl)cc24)C1([2H])[2H] |
| IUPAC Name | tert-butyl 4-(8-chloro-3-methoxybenzo[b][1,4]benzoxazepin-6-yl)-2,2,3,3,5,5,6,6-octadeuteriopiperazine-1-carboxylate |
| Molecular Weight (g/mol) | 451.973 |
| Formula Weight | 451.211 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction