missing translation for 'onlineSavingsMsg'
Learn More

n-Tetradecylphosphocholine, CDN

Product Code. 30313154
Change view
Click to view available options
Quantity:
0.1 g
Unit Size:
0.1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313154 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313154 Supplier CDN Supplier No. CDNX58860.1G
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetradecylphosphocholine
CAS 77733-28-9
Molecular Formula C19 H42 N O4 P
Quantity 0.1 g
Synonym Ethanaminium, 2-[[hydroxy(tetradecyloxy)phosphinyl]oxy]-N,N,N-trimethyl-, inner salt (9 CI, ACI), D 19390, FOS 14, Fos-choline 14, n-Tetradecylphosphocholine, Tetradecylphosphocholine, Tetradecylphosphorylcholine, O-(Tetradecylphosphoryl)choline, 2-[[Hydroxy(tetradecyloxy)phosphinyl]oxy]-N,N,N-trimethylethanaminium inner salt
InChI Formula InChI=1 S/C19H42NO4P/c1-5-6-7-8-9-10-11-12-13-14-15-16-18-23-25(21,22)24-19-17-20(2,3)4/h5-19H2,1-4H3
SMILES CCCCCCCCCCCCCCOP(=O)([O-])OCC[N+](C)(C)C
IUPAC Name tetradecyl 2-(trimethylazaniumyl)ethyl phosphate
Molecular Weight (g/mol) 379.515
Formula Weight 379.285 g/mol
Percent Purity min 96% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.