missing translation for 'onlineSavingsMsg'
Learn More

N-Triphenylmethyl-5-[4'-methylbiphenyl-2-yl]tetrazole, TRC

Product Code. 30454841
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454841 250 mg 250mg
30450964 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454841 Supplier Toronto Research Chemicals Supplier No. TRCT889170250MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133909-97-4
Molecular Formula C33 H26 N4
Molecular Weight (g/mol) 478.59
Synonym 2H-Tetrazole, 5-(4'-methyl[1,1'-biphenyl]-2-yl)-2-(triphenylmethyl)-, 5-(4'-Methyl[1,1'-biphenyl]-2-yl)-2-(triphenylmethyl)-2H-tetrazole
IUPAC Name 5-[2-(4-methylphenyl)phenyl]-2-trityltetrazole
SMILES Cc1ccc(cc1)c2ccccc2c3nnn(n3)C(c4ccccc4)(c5ccccc5)c6ccccc6

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C33H26N4/c1-25-21-23-26(24-22-25)30-19-11-12-20-31(30)32-34-36-37(35-32)33(27-13-5-2-6-14-27,28-15-7-3-8-16-28)29-17-9-4-10-18-29/h2-24H,1H3
Formula Weight 478.2157
Chemical Name or Material N-Triphenylmethyl-5-[4'-methylbiphenyl-2-yl]tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.