missing translation for 'onlineSavingsMsg'
Learn More

N2,N2,N6-Tris[(1,1-dimethylethoxy)carbonyl]-L-lysine 1,1-Dimethylethylester, TRC

Product Code. 30456458
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456458 250 mg 250mg
30457082 500 mg 500mg
30455305 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456458 Supplier Toronto Research Chemicals Supplier No. TRCT886525250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N2,N2,N6-Tris[(1,1-dimethylethoxy)carbonyl]-L-lysine 1,1-Dimethylethylester
Analyte or Component Names N2,N2,N6-Tris[(1,1-dimethylethoxy)carbonyl]-L-lysine 1,1-Dimethylethylester
Shipping Condition Room Temperature
Molecular Formula C25 H46 N2 O8
InChI Formula InChI=1S/C25H46N2O8/c1-22(2,3)32-18(28)17(15-13-14-16-26-19(29)33-23(4,5)6)27(20(30)34-24(7,8)9)21(31)35-25(10,11)12/h17H,13-16H2,1-12H3,(H,26,29)
Solution Type Neat
SMILES CC(C)(C)OC(=O)NCCCCC(N(C(=O)OC(C)(C)C)C(=O)OC(C)(C)C)C(=O)OC(C)(C)C
IUPAC Name tert-butyl 2-[bis[(2-methylpropan-2-yl)oxycarbonyl]amino]-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 502.641
Quantity 250 mg
Formula Weight 502.325
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.