missing translation for 'onlineSavingsMsg'
Learn More

N8-Acetylspermidine Dihydrochloride, TRC

Product Code. 30006659
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006659 50 mg 50mg
30006234 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006659 Supplier Toronto Research Chemicals Supplier No. TRCA18800050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34450-15-2
Molecular Formula C9H23Cl2N3O
Molecular Weight (g/mol) 260.2
Synonym N-[4-[(3-Aminopropyl)amino]butyl]acetamide Dihydrochloride, N-[4-[(3-Aminopropyl)amino]butyl]acetamide Hydrochloride (1:2)
IUPAC Name N-[4-(3-aminopropylamino)butyl]acetamide
SMILES NCCCNCCCCNC(C)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H21N3O/c1-9(13)12-8-3-2-6-11-7-4-5-10/h11H,2-8,10H2,1H3,(H,12,13)
Formula Weight 187.1685
Percent Purity >95%
Chemical Name or Material N8-Acetylspermidine Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.