missing translation for 'onlineSavingsMsg'
Learn More

Nadolol, TRC

Product Code. 18041110
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18041110 50 mg 50mg
18062310 1 g 1g
18030820 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18041110 Supplier Toronto Research Chemicals Supplier No. TRCN20105050MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42200-33-9
Molecular Formula C17 H27 N O4
Molecular Weight (g/mol) 309.4
Synonym 5-[3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-1,2,3,4-tetrahydro-2,3-naphthalenediol, Anabet, Corgard, Nadolol, SQ 11725, Solgol, cis-5-[(2RS)-3-[(1,1-Dimethylethyl)amino]-2-hydroxypropoxy]-1,2,3,4-tetrahydronaphthalene-2,3-diol, 1-(tert-Butylamino)-3-[(5,6,7,8-tetrahydro-cis-6,7-dihydroxy-1-naphthyl)oxy]-2-propanol, 2,3-cis-1,2,3,4-tetrahydro-5-[2-hydroxy-3-(tert-butylamino)propoxy]-2,3-naphthalenediol
IUPAC Name (2R,3S)-5-[3-(tert-butylamino)-2-hydroxypropoxy]-1,2,3,4-tetrahydronaphthalene-2,3-diol
SMILES CC(C)(C)NCC(O)COc1cccc2C[C@@H](O)[C@@H](O)Cc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H27NO4/c1-17(2,3)18-9-12(19)10-22-16-6-4-5-11-7-14(20)15(21)8-13(11)16/h4-6,12,14-15,18-21H,7-10H2,1-3H3/t12?,14-,15+/m1/s1
Formula Weight 309.194
Chemical Name or Material Nadolol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.