missing translation for 'onlineSavingsMsg'
Learn More

Melford NADPH

Product Code. 30018569
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018569 100 mg 100mg
30018571 1 g 1g
30018570 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018569 Supplier Melford Supplier No. N201400.1
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • Substance Name: β-Nicotinamide Adenine Dinucleotide Phosphate, Reduced Form, Tetra Sodium Salt Trihydrate
  • CAS Number: 2646-71-1
  • Molecular Formula: C21H26N7O17 P3Na4 3H2O

Specifications

Quantity 100 mg
Molecular Formula C21H26N7O17P3Na4-3H2O
CAS 2646-71-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.