missing translation for 'onlineSavingsMsg'
Learn More

Nafamostat Mesylate, TRC

Product Code. 30005509
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005509 50 mg 50mg
30006528 10 mg 10mg
30004521 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005509 Supplier Toronto Research Chemicals Supplier No. TRCN21000050MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82956-11-4
Molecular Formula C19 H17 N5 O2 . 2 C H4 O3 S
Molecular Weight (g/mol) 539.58
IUPAC Name (6-carbamimidoylnaphthalen-2-yl) 4-carbamimidamidobenzoate;methanesulfonic acid
SMILES CS(=O)(=O)O.CS(=O)(=O)O.NC(=N)Nc1ccc(cc1)C(=O)Oc2ccc3cc(ccc3c2)C(=N)N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H17N5O2.2CH4O3S/c20-17(21)14-2-1-13-10-16(8-5-12(13)9-14)26-18(25)11-3-6-15(7-4-11)24-19(22)23;2*1-5(2,3)4/h1-10H,(H3,20,21)(H4,22,23,24);2*1H3,(H,2,3,4)
Formula Weight 539.1145
Chemical Name or Material Nafamostat, Mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.