Learn More
Thermo Scientific™ Nalgene™ Stainless Steel, Handle
Product Code. 10623923
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10623923 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10623923 Supplier Thermo Scientific™ Supplier No. 22290001
  • ONLINE EXCLUSIVE PRICE
    46.40€
    42.90€ / Pack of 1
    Save 3.50€
    7% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For Use With: 20 L Single Use Carboys
Easily transport fluids with the Thermo Scientific™ Nalgene™ Handle for Single-Use Carboy, a stainless-steel handle that attaches to 20 L carboys. For use with Nalgene™ Sterile Single Use Carboy (Cat. No. 342889-0050).
  • Stainless-steel handle
  • For use with Nalgene™ Sterile Single Use Carboy (Cat. No. 342889-0050)
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
For Use With (Equipment) Carboys, 20 L
Material Stainless Steel
No. per Case 1 Ea.
Product Type Handle
Quantity Each
Workflow Step Upstream process solution storage. Upstream Validation