missing translation for 'onlineSavingsMsg'
Learn More

NAlpha-Benzoyl-DL-arginine Beta-Naphthylamide Hydrochloride, TRC

Product Code. 30133389
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133389 50 mg 50mg
30135008 1 g 1g
30004941 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133389 Supplier Toronto Research Chemicals Supplier No. TRCB20790850MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913-04-2
Molecular Formula C23H25N5O2.HCl
Molecular Weight (g/mol) 439.9
Synonym BANA, N-Benzoyl-DL-arginine-beta-naphthylamide Hydrochloride, N-alpha-Benzoyl-DL-arginine-beta-naphthylamide Hydrochloride, NSC 83266, alpha-N-Benzoyl-DL-arginine-2-naphthylamide Hydrochloride, alpha-N-Benzoyl-DL-arginine-beta-naphthylamide Hydrochloride, (±)-N-[4-Guanidino-1-(2-naphthylcarbamoyl)butyl]-benzamide Monohydrochloride, (±)-N-[4-[(Aminoiminomethyl)amino]-1-[(2-naphthalenylamino)carbonyl]butyl]-benzamide Monohydrochloride;
IUPAC Name N-[5-(diaminomethylideneamino)-1-(naphthalen-2-ylamino)-1-oxopentan-2-yl]benzamide;hydrochloride
SMILES Cl.NC(=N)NCCCC(NC(=O)c1ccccc1)C(=O)Nc2ccc3ccccc3c2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C23H25N5O2.ClH/c24-23(25)26-14-6-11-20(28-21(29)17-8-2-1-3-9-17)22(30)27-19-13-12-16-7-4-5-10-18(16)15-19;/h1-5,7-10,12-13,15,20H,6,11,14H2,(H,27,30)(H,28,29)(H4,24,25,26);1H
Formula Weight 439.1775
Chemical Name or Material Nalpha-Benzoyl-DL-arginine beta-Naphthylamide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.