Learn More
Thermo Scientific™ NanoDrop™ Eight Tube Rack
Product Code. 31921084 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
NanoDrop Eight Spectrophotometers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31921084 NanoDrop Eight Spectrophotometers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 31921084 Supplier Thermo Scientific™ Supplier No. 840343700
 Restricted Item
In Stock
Lägg i varukorgen
Lägg i varukorgen
missing translation for 'validMainframeMsg'
Denna artikel kan inte returneras. Se returpolicy
Regleringsinformation: Denna produkt innehåller ett ämne som är upptaget i bilaga XIV till REACH-förordningen (EG) nr 1907/2006. Alla kunder måste fylla i en deklaration före leverans.
The Thermo Scientific™ NanoDrop™ Eight Tube Rack allows you to place samples in tubes on the instrument LED board to help you keep track of samples measured.
The Thermo Scientific™ NanoDrop™ Eight Tube Rack allows you to place samples in tubes on the instrument LED board to help you keep track of samples measured.
This single tube rack allows users to keep track of sample measurements for samples stored in 0.5 mL and 1.5 mL-sized tubes.
Specifikationer
Description NanoDrop Eight Tube Rack
For Use With (Equipment) NanoDrop Eight Spectrophotometers
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.