missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-2-sulfonic Acid Hydrate, TRC

Product Code. 30473124
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473124 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473124 Supplier Toronto Research Chemicals Supplier No. TRCN3785035G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Naphthylsulfonic Acid
CAS 120-18-3
Molecular Formula C10 H8 O3 S
Quantity 5 g
Synonym 2-Naphthylsulfonic acid, 2-Sulfonaphthalene, Dirinal NSK, β-Naphthalenesulfonic acid, β-Naphthylsulfonic acid
InChI Formula InChI=1S/C10H8O3S/c11-14(12,13)10-6-5-8-3-1-2-4-9(8)7-10/h1-7H,(H,11,12,13)
SMILES OS(=O)(=O)c1ccc2ccccc2c1
IUPAC Name naphthalene-2-sulfonic acid
Molecular Weight (g/mol) 208.234
Formula Weight 208.019
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.