Learn More
Naphthalene-2-sulfonohydrazide, TRC
Product Code. 30428651
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428651 500 mg 500mg
30408878 100 mg -
30417694 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30428651 Supplier Toronto Research Chemicals Supplier No. TRCN756615500MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10151-46-9
Molecular Formula C10 H10 N2 O2 S
Molecular Weight (g/mol) 222.264
Synonym (Naphthalen-2-yl)sulfonyl Hydrazine, (beta-Naphthylsulfonyl)hydrazine, 2-Naphthalenesulfonyl Hydrazide, 2-Naphthylsulfonyl Hydrazide2-Naphthylsulfonyl Hydrazine
IUPAC Name naphthalene-2-sulfonohydrazide
SMILES NNS(=O)(=O)c1ccc2ccccc2c1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C10H10N2O2S/c11-12-15(13,14)10-6-5-8-3-1-2-4-9(8)7-10/h1-7,12H,11H2
Formula Weight 222.046
Chemical Name or Material Naphthalene-2-sulfonohydrazide