missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene, Extra Pure, SLR, Fisher Chemical

Product Code. 10234060 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10234060 500 g Glass Jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10234060 Supplier Fisher Chemical Supplier No. N/0150/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 91-20-3
Melting Point 79°C
Color White
Boiling Point 218°C
Packaging Glass Jar
Molecular Formula C10H8
MDL Number MFCD00001742
Quantity 500 g
Synonym naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene
InChI Key UFWIBTONFRDIAS-UHFFFAOYSA-N
SMILES C1=CC2=CC=CC=C2C=C1
IUPAC Name naphthalene
Molecular Weight (g/mol) 128.17
PubChem CID 931
ChEBI CHEBI:16482
Formula Weight 128.17g/mol
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Naphthalene
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.