missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Napropamide-d10 (N,N-diethyl-d10), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (±)-Napropamide-d10 (N,N-diethyl-d10) |
| Molecular Formula | C17H11D10NO2 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C17H21NO2/c1-4-18(5-2)17(19)13(3)20-16-12-8-10-14-9-6-7-11-15(14)16/h6-13 H,4-5H2,1-3H3/i1D3,2D3,4D2,5D2 |
| SMILES | O=C(N(C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])C(C)OC1=CC=CC2=CC=CC=C21 |
| IUPAC Name | N,N-bis(ethyl-d5)-2-(naphthalen-1-yloxy)propanamide |
| Molecular Weight (g/mol) | 281.42 |
| Formula Weight | 281.22 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction