missing translation for 'onlineSavingsMsg'
Learn More

Naproxen-d3, TRC

Product Code. 30474146
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474146 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30474146 Supplier Toronto Research Chemicals Supplier No. TRCN37752425MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naproxen-d3
CAS 958293-79-3
Purity Grade Notes HPLC
Molecular Formula C14H11D3O3
Quantity 25 mg
Synonym 2-Naphthaleneacetic acid','6-(methoxy-d3)-α-methyl-, 6-(Methoxy-d3)-α-methyl-2-naphthaleneacetic acid, 2-[6-(Trideuteromethoxy)naphthalen-2-yl]propanoic acid, rac-Naproxen (methoxy-D3), (R,S)-Naproxen D3 (methoxy D3)
InChI Formula InChI=1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/i2D3
SMILES [2H]C([2H])([2H])Oc1ccc2cc(ccc2c1)C(C)C(=O)O
IUPAC Name 2-[6-(trideuteriomethoxy)naphthalen-2-yl]propanoic acid
Molecular Weight (g/mol) 233.28
Formula Weight 233.1131
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.