missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAT13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16120-1-AP-20UL
This item is not returnable.
View return policy
Description
The XPA (xeroderma pigmentosumgroup A) protein specifically recognizes the UV-orchemically damaged DNA lesions, and triggers thenucleotide excision repair process. XPA binds to thereplication protein A (RPA) or the excision repaircross complementing 1 protein (ERCC 1). In the absence of nucleotide excision repair persisting(unrepaired) DNA lesions (adducts) may lead to the accumulation of gene mutations and ultimately to cancer. Xeroderma pigmentosum patients have a >2000 fold increased risk to develop skin cancer atsun-exposed areas.Specifications
| NAT13 | |
| Polyclonal | |
| Unconjugated | |
| NAA50 | |
| hNAT5, hSAN, MAK3, N acetyltransferase 13, N acetyltransferase 5, NAA50, NAT13, NAT5, SAN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288108, 72117, 80218 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PGB6, Q9GZZ1 | |
| Naa50 | |
| NAT13 Fusion Protein Ag9075 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction