Learn More
Piercan™ Natural Rubber Latex Chemical Resistant Gloves
Product Code. 11799528
Change view
Click to view available options
Size:
8
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11799528 8 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11799528 Supplier Piercan™ Supplier No. 10800H4/10T8
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Certifications/Compliance ISO 9001 and ISO 14001
Coating Material Natural Rubber Latex
Hand Ambidextrous
Latex Free Yes
Material Rubber Latex
Product Type Gloves
Size 8
Thickness (Metric) 0.4 mm
Quantity 1 Pair
Resistant To Alkalis, Aliphatic solvents and Ketones
Length (Metric) 800 mm