Learn More
Abeba™ Nature 4090 Shoes
Leather covered cork footbed is ESD dissipative.
Brand:  Abeba™ 4090/41
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15641478
  • 81.95€ / 1 pair
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • 2 adjustable instep straps
  • velcro fastening
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Cork
41
Rubber
White
Leather
Slip-resistant
4090