missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nav1.7 Antibody (Duke anti-NAv1.7), mFluor Violet 500 SE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27987MFV500
This item is not returnable.
View return policy
Description
Nav1.7 Monoclonal antibody specifically detects Nav1.7 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Nav1.7 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| ETHA, FEB3B, NE-NA, NENAVoltage-gated sodium channel subunit alpha Nav1.7, Neuroendocrine sodium channel, Peripheral sodium channel 1, PN1hNE-Na, sodium channel protein type 9 subunit alpha, Sodium channel protein type IX subunit alpha, sodium channel, voltage-gated, type IX, alpha subunit, voltage-gated sodium channel alpha subunit Nav1.7, voltage-gated, type IX, alpha polypeptide | |
| SCN9a/ Nav1.7 | |
| 0.1 mL | |
| Cancer, Cellular Signaling, Neuroscience | |
| 6335 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Duke anti-NAv1.7 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction