missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15445-1-AP-150UL
This item is not returnable.
View return policy
Description
NDUFA6 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6), also known as LYRM6 (LYR motif-containing protein 6), NADHB14 or B14, is a 128 amino acid protein that localizes to the matrix side of the mitochondrial membrane and belongs to the complex I LYR family. Existing as a subunit of the multi-protein membrane respiratory chain NADH dehydrogenase complex (Complex I), NDUFA6 functions as an accessory protein that facilitates the transfer of electrons from NADH to the respiratory chain. The gene encoding NDUFA6 maps to human chromosome 22, which houses over 500 genes and is the second smallest human chromosome.Specifications
| NDUFA6 | |
| Polyclonal | |
| Unconjugated | |
| NDUFA6 | |
| B14, CI B14, Complex I B14, LYR motif containing protein 6, LYRM6, NADHB14, NDUFA6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4700 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P56556 | |
| NDUFA6 | |
| NDUFA6 Fusion Protein Ag7593 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction